Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001484.2 | NP_001001484.1 | 6 | 2311 | ||
NM_001261836.1 | NP_001248765.1 | 5 | 2177 | |||
NM_001261837.1 | NP_001248766.1 | 5 | 2170 | |||
NM_001261838.1 | NP_001248767.1 | 4 | 1697 | |||
NM_030664.4 | NP_109589.2 | 6 | 2256 | |||
XM_017016927.1 | XP_016872416.1 | 5 | 2095 | |||
XM_017016928.1 | XP_016872417.1 | 4 | 2130 | |||
XM_017016929.1 | XP_016872418.1 | 4 | 2036 | |||
XM_017016930.1 | XP_016872419.1 | 4 | 2007 | |||
GenBank mRNA | AL119009.1 | - | 1 | 268 | ||
BC015092.1 | - | 6 | 2234 | 4049884 | ||
BC050411.1 | - | 6 | 2283 | 6472327 | ||
BX648556.1 | - | 5 | 2149 |