Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTDSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030783.1 | NP_110410.1 | 8, 9 | 1039 | ||
XM_005253153.1 | XP_005253210.1 | 8, 9 | 1020 | |||
XM_011520391.2 | XP_011518693.1 | 8, 9 | 1657 | |||
XM_011520392.2 | XP_011518694.1 | 9, 10 | 1036 | |||
XM_011520393.2 | XP_011518695.1 | 7, 8 | 754 | |||
XM_017018368.1 | XP_016873857.1 | 10, 11 | 3431 | |||
XM_017018369.1 | XP_016873858.1 | 9, 10 | 1161 | |||
XM_017018370.1 | XP_016873859.1 | 7, 8 | 836 | |||
XM_017018371.1 | XP_016873860.1 | 8, 9 | 950 | |||
GenBank mRNA | AL834357.1 | - | 8, 9 | 953 | ||
BC001210.1 | - | 8, 9 | 1039 | 3503123 |