Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030785.3 | NP_110412.1 | 6 | 2364 | ||
GenBank mRNA | AK093068.1 | - | 5 | 1634 | ||
AK098837.1 | - | 6 | 2309 | |||
AK223347.1 | - | 6 | 2309 | |||
AL136761.1 | - | 6 | 2336 | |||
BC057785.1 | - | 6 | 2192 | |||
BC136351.1 | - | 6 | 2245 | 9020338 | ||
BC136352.1 | - | 6 | 2245 | 9020339 |