Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281441.1 | NP_001268370.1 | 3, 4 | 727 | ||
NM_030798.4 | NP_110425.2 | 3, 4 | 727 | |||
NM_148842.2 | NP_683682.1 | 3, 4 | 727 | |||
GenBank mRNA | AF410455.1 | - | 3, 4 | 583 | ||
AK057870.1 | - | 966 | ||||
AK309117.1 | - | 3, 4 | 636 | |||
AL136804.1 | - | 3, 4 | 634 | |||
AL536085.3 | - | 3, 4 | 725 | |||
BC007823.2 | - | 3, 4 | 639 | 4304040 | ||
BC019008.2 | - | 3, 4 | 639 | 4330507 | ||
BC032712.1 | - | 3, 4 | 658 | 5495794 | ||
BC040695.1 | - | 3, 4 | 709 | 5785681 |