Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025579.2 | NP_001020750.1 | 3 | 341 | ||
NM_030808.4 | NP_110435.1 | 3 | 341 | |||
XM_011524013.1 | XP_011522315.1 | 3 | 332 | |||
XM_017025183.1 | XP_016880672.1 | 4 | 521 | |||
XM_017025184.1 | XP_016880673.1 | 4 | 521 | |||
XM_017025185.1 | XP_016880674.1 | 3 | 390 | |||
XM_017025186.1 | XP_016880675.1 | 4 | 521 | |||
XM_017025187.1 | XP_016880676.1 | 3 | 390 | |||
XM_017025188.1 | XP_016880677.1 | 4 | 521 | |||
GenBank mRNA | AF182078.1 | - | 3 | 268 | ||
AF217798.3 | - | 3 | 332 | |||
AK056014.1 | - | 3 | 339 | |||
AK299559.1 | - | 4 | 315 | |||
AL832648.1 | - | 3 | 275 | |||
AY004871.1 | - | 3 | 267 | |||
BC026101.2 | - | 3 | 279 | 4820855 | ||
BP201698.1 | - | 3 | 341 | |||
CX166766.1 | - | 3 | 270 | |||
DB176966.1 | - | 3 | 341 | |||
DN993952.1 | - | 3 | 332 |