Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030816.4 | NP_110443.3 | 1 | 711 | ||
XM_005271234.3 | XP_005271291.1 | 1 | 954 | |||
XM_005271235.3 | XP_005271292.1 | 1 | 701 | |||
XM_006710929.3 | XP_006710992.1 | 1 | 724 | |||
XM_017002414.1 | XP_016857903.1 | 1 | 62 | |||
XM_017002415.1 | XP_016857904.1 | 1 | 62 | |||
GenBank mRNA | AL136717.1 | - | 1 | 712 | ||
BC016937.2 | - | 1 | 693 | |||
BC028840.1 | - | 1 | 700 | 4558748 |