Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320561.1 | NP_001307490.1 | 7, 8 | 898 | ||
NM_001320564.1 | NP_001307493.1 | 7, 8 | 886 | |||
NM_001320565.1 | NP_001307494.1 | 7, 8 | 768 | |||
NM_001320566.1 | NP_001307495.1 | 7, 8 | 886 | |||
NM_001320567.1 | NP_001307496.1 | 7, 8 | 749 | |||
NM_001320568.1 | NP_001307497.1 | 6, 7 | 558 | |||
NM_001320569.1 | NP_001307498.1 | 5, 6 | 354 | |||
NM_030818.3 | NP_110445.1 | 6, 7 | 558 | |||
XM_005260086.4 | XP_005260143.1 | 7, 8 | 748 | |||
XM_011528326.1 | XP_011526628.1 | 6, 7 | 637 | |||
GenBank mRNA | AF250306.1 | - | 6, 7 | 486 | ||
AK098262.1 | - | 6, 7 | 515 | |||
AK293927.1 | - | 7, 8 | 749 | |||
AL560878.3 | - | 7, 8 | 769 | |||
BC002905.2 | - | 6, 7 | 482 | 3940130 | ||
BI913255.1 | - | 5, 6 | 319 | |||
BM547551.1 | - | 6, 7 | 556 | |||
BX394050.2 | - | 7, 8 | 879 | |||
BX422326.2 | - | 6, 7 | 557 | |||
CR999732.1 | - | 5, 6 | 483 |