Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181900.2 | NP_871629.1 | 6 | 1067 | ||
NR_135013.1 | 5 | 1017 | ||||
XM_017022639.1 | XP_016878128.1 | 5 | 1582 | |||
GenBank mRNA | AK026352.1 | - | 5 | 975 | ||
AK097024.1 | - | 4 | 2435 | |||
AK225479.1 | - | 5 | 975 | |||
AL137657.1 | - | 5 | 1017 | |||
BC004365.1 | - | 6 | 1007 | 3641640 |