Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136026.2 | NP_001129498.1 | 10 | 1502 | ||
NM_001136232.2 | NP_001129704.1 | 9 | 1356 | |||
NM_030673.3 | NP_109598.2 | 10 | 1358 | |||
NM_183352.2 | NP_899195.1 | 9 | 1324 | |||
XM_005265379.2 | XP_005265436.1 | 10 | 1375 | |||
XM_017007019.1 | XP_016862508.1 | 10 | 1412 | |||
XM_017007020.1 | XP_016862509.1 | 9 | 992 | |||
XM_017007021.1 | XP_016862510.1 | 9 | 1002 | |||
GenBank mRNA | AB209554.1 | - | 9 | 3498 | ||
AF052155.1 | - | 9 | 1356 | |||
AK095629.1 | - | 7 | 3183 | |||
BC002634.2 | - | 10 | 1239 | 3605545 | ||
BC006167.1 | - | 6 | 1016 | |||
BC091506.1 | - | 9 | 1234 | 30408210 | ||
DB564012.1 | - | 1 | 300 | |||
L09260.1 | - | 9 | 1201 |