Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030753.4 | NP_110380.1 | 5 | 1522 | ||
GenBank mRNA | BC114219.1 | - | 5 | 1377 | 5726751 |