Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282741.1 | NP_001269670.1 | 4, 5 | 759 | ||
NM_030758.3 | NP_110385.1 | 5, 6 | 1522 | |||
XM_005261465.4 | XP_005261522.1 | 5, 6 | 1448 | |||
XM_006724207.2 | XP_006724270.1 | 4, 5 | 705 | |||
XM_006724208.3 | XP_006724271.1 | 3, 4 | 490 | |||
XM_011530057.1 | XP_011528359.1 | 5, 6 | 1448 | |||
XM_011530059.2 | XP_011528361.1 | 5, 6 | 1448 | |||
XM_011530060.1 | XP_011528362.1 | 4, 5 | 759 | |||
XM_011530061.1 | XP_011528363.1 | 3, 4 | 433 | |||
XM_011530062.1 | XP_011528364.1 | 5, 6 | 804 | |||
XM_017028708.1 | XP_016884197.1 | 5, 6 | 1448 | |||
XM_017028710.1 | XP_016884199.1 | 5, 6 | 911 | |||
XM_017028711.1 | XP_016884200.1 | 4, 5 | 1209 | |||
GenBank mRNA | AB051451.2 | - | 5, 6 | 1430 | ||
AF288741.1 | - | 5, 6 | 1416 | |||
AF323731.1 | - | 5, 6 | 1522 | |||
AK131374.1 | - | 5, 6 | 1049 | |||
AK294007.1 | - | 5, 6 | 1454 | |||
AK296354.1 | - | 4, 5 | 759 | |||
BC004340.1 | - | 2, 3 | 317 |