Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030780.4 | NP_110407.2 | 7 | 1604 | ||
NR_102337.1 | 6 | 1518 | ||||
NR_102338.1 | 8 | 1713 | ||||
XM_017013877.1 | XP_016869366.1 | 7 | 1507 | |||
GenBank mRNA | AF283645.1 | - | 7 | 1334 | ||
AK027787.1 | - | 7 | 1337 | |||
AK125492.1 | - | 8 | 1321 | |||
BC021893.1 | - | 7 | 1311 | 4524907 |