Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VOPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284282.1 | NP_001271211.1 | 5 | 1364 | ||
NM_001284283.1 | NP_001271212.1 | 5 | 1298 | |||
NM_001284284.1 | NP_001271213.1 | 5 | 1459 | |||
NM_001321242.1 | NP_001308171.1 | 5 | 1379 | |||
NM_001321243.1 | NP_001308172.1 | 6 | 1532 | |||
NM_001321244.1 | NP_001308173.1 | 6 | 1565 | |||
NM_001321246.1 | NP_001308175.1 | 4 | 1415 | |||
NM_001321247.1 | NP_001308176.1 | 4 | 1299 | |||
NM_001321248.1 | NP_001308177.1 | 4 | 1242 | |||
NM_001321249.1 | NP_001308178.1 | 5 | 1381 | |||
NM_001321250.1 | NP_001308179.1 | 5 | 1403 | |||
NM_001321251.1 | NP_001308180.1 | 5 | 1389 | |||
NM_030796.4 | NP_110423.3 | 5 | 1493 | |||
GenBank mRNA | AB097004.1 | - | 5 | 1352 | ||
AF395824.1 | - | 4 | 1258 | |||
AK075391.1 | - | 6 | 1427 | |||
AK090825.1 | - | 5 | 1403 | |||
AK092992.1 | - | 5 | 1437 | |||
AK126848.1 | - | 1 | 1794 | |||
AK315882.1 | - | 5 | 1291 | |||
AL136610.1 | - | 1 | 1306 | |||
BC016650.1 | - | 1 | 1385 | 4342536 |