Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL21A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318751.1 | NP_001305680.1 | 24 | 2415 | ||
NM_001318752.1 | NP_001305681.1 | 22 | 2296 | |||
NM_001318753.1 | NP_001305682.1 | 7 | 462 | |||
NM_001318754.1 | NP_001305683.1 | 7 | 462 | |||
NM_030820.3 | NP_110447.2 | 23 | 2341 | |||
NR_134849.1 | 23 | 2333 | ||||
NR_134850.1 | 24 | 2358 | ||||
XM_011514924.2 | XP_011513226.1 | 23 | 2284 | |||
XM_011514925.2 | XP_011513227.1 | 25 | 2454 | |||
XM_011514926.1 | XP_011513228.1 | 23 | 2212 | |||
XM_011514927.1 | XP_011513229.1 | 23 | 2166 | |||
GenBank mRNA | AF330693.1 | - | 22 | 2262 | ||
AF370383.1 | - | 11 | 597 | |||
AF414088.1 | - | 23 | 2311 | |||
AK096444.1 | - | 7 | 465 | |||
AK126586.1 | - | 24 | 2226 | |||
AK313398.1 | - | 23 | 2317 | |||
AL136624.1 | - | 23 | 2311 | |||
BC045597.1 | - | 23 | 2288 | 4814199 | ||
BC126108.1 | - | 23 | 2196 | 8991824 | ||
BC143862.1 | - | 23 | 2188 | |||
BC143863.1 | - | 24 | 2213 | |||
BC143865.1 | - | 24 | 2270 | 9052381 | ||
DA826579.1 | - | 7 | 452 |