Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030923.4 | NP_112185.1 | 1, 2 | 269 | ||
XM_017005060.1 | XP_016860549.1 | 1, 2 | 356 | |||
XM_017005062.1 | XP_016860551.1 | 1, 2 | 334 | |||
GenBank mRNA | BC026170.2 | - | 1, 2 | 269 | 4837567 |