Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042517.1 | NP_001035982.1 | 10 | 1300 | ||
NM_001258366.1 | NP_001245295.1 | 9 | 1267 | |||
NM_001258367.1 | NP_001245296.1 | 8 | 1162 | |||
NM_001258368.1 | NP_001245297.1 | 8 | 1090 | |||
NM_001258369.1 | NP_001245298.1 | 10 | 1300 | |||
NM_001258370.1 | NP_001245299.1 | 4 | 471 | |||
NM_030932.3 | NP_112194.2 | 4 | 471 | |||
XM_006719876.1 | XP_006719939.1 | 4 | 451 | |||
XM_011535258.2 | XP_011533560.1 | 10 | 1243 | |||
XM_011535263.2 | XP_011533565.1 | 10 | 1243 | |||
XM_011535265.2 | XP_011533567.1 | 10 | 1243 | |||
XM_017020789.1 | XP_016876278.1 | 10 | 1243 | |||
GenBank mRNA | AB244756.1 | - | 9 | 1048 | ||
AB244757.1 | - | 8 | 943 | |||
AB244758.1 | - | 8 | 871 | |||
AK092024.1 | - | 11 | 1222 | |||
AK298509.1 | - | 10 | 1086 | |||
AK307652.1 | - | 4 | 451 | |||
AK316474.1 | - | 10 | 1213 | |||
AL137718.1 | - | 4 | 443 | |||
AY750055.1 | - | 10 | 1173 | |||
AY818645.1 | - | 10 | 1081 | |||
BC034952.1 | - | 4 | 475 | 4830888 | ||
BC048963.1 | - | 4 | 427 | 5265490 | ||
BC068504.1 | - | 4 | 427 | 4838507 | ||
BX649186.1 | - | 10 | 1242 |