Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320153.1 | NP_001307082.1 | 8 | 1586 | ||
NM_001320155.1 | NP_001307084.1 | 8 | 876 | |||
NM_030937.4 | NP_112199.2 | 7 | 862 | |||
NR_135154.1 | 7 | 2800 | ||||
XM_011542216.2 | XP_011540518.1 | 7 | 937 | |||
XM_011542219.2 | XP_011540521.1 | 7 | 746 | |||
XM_011542221.2 | XP_011540523.1 | 6 | 577 | |||
XM_017002420.1 | XP_016857909.1 | 8 | 2367 | |||
XM_017002421.1 | XP_016857910.1 | 9 | 1250 | |||
XM_017002422.1 | XP_016857911.1 | 8 | 984 | |||
GenBank mRNA | AF251294.1 | - | 6 | 1315 | ||
AK000685.1 | - | 2 | 402 | |||
AK026796.1 | - | 2 | 1061 | |||
AK027770.1 | - | 2 | 1831 | |||
AK056120.1 | - | 8 | 1558 | |||
AK057133.1 | - | 6 | 577 | |||
AK074112.1 | - | 7 | 2771 | |||
AK225758.1 | - | 7 | 857 | |||
AK292268.1 | - | 2 | 385 | |||
AK293451.1 | - | 4 | 340 | |||
AY037150.1 | - | 7 | 836 | |||
AY116620.1 | - | 8 | 865 | |||
BC071622.1 | - | 2 | 249 |