Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030949.2 | NP_112211.1 | 1 | 356 | ||
GenBank mRNA | AF308297.1 | - | 1 | 229 | ||
AF407165.2 | - | 1 | 858 | |||
BC015773.1 | - | 1 | 332 | 4877511 | ||
CB994599.1 | - | 1 | 384 | |||
DR003369.1 | - | 1 | 366 |