Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160223.1 | NP_001153695.1 | 3 | 656 | ||
NM_001160224.1 | NP_001153696.1 | 3 | 426 | |||
NM_001160225.1 | NP_001153697.1 | 3 | 305 | |||
NM_030954.3 | NP_112216.3 | 3 | 426 | |||
NR_027668.1 | 3 | 656 | ||||
NR_027669.1 | 3 | 426 | ||||
XM_006716404.2 | XP_006716467.1 | 4 | 413 | |||
XM_011544666.2 | XP_011542968.1 | 3 | 210 | |||
XM_017013881.1 | XP_016869370.1 | 4 | 353 | |||
GenBank mRNA | AK027748.1 | - | 3 | 342 | ||
AK090864.1 | - | 3 | 589 | |||
AK225086.1 | - | 3 | 306 | |||
AK225568.1 | - | 3 | 322 | |||
AL136620.1 | - | 3 | 310 | |||
BC013422.1 | - | 3 | 291 | |||
BC032393.1 | - | 3 | 288 | 5200847 | ||
BC039461.1 | - | 3 | 413 | |||
BC044566.1 | - | 3 | 426 | |||
BC058289.1 | - | 3 | 289 | |||
BG910500.1 | - | 3 | 647 | |||
DA751961.1 | - | 3 | 426 |