Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030957.3 | NP_112219.3 | 5 | 773 | ||
XM_006722917.3 | XP_006722980.1 | 4 | 614 | |||
XM_017027338.1 | XP_016882827.1 | 4 | 629 | |||
GenBank mRNA | AB209515.1 | - | 4 | 604 | ||
AF163762.1 | - | 2 | 389 | |||
BX461830.2 | - | 4 | 551 |