Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146008.1 | NP_001139480.1 | 1 | 826 | ||
NM_001146009.1 | NP_001139481.1 | 1 | 605 | |||
NM_030958.2 | NP_112220.2 | 2 | 816 | |||
XM_005251313.2 | XP_005251370.1 | 2 | 816 | |||
XM_017013883.1 | XP_016869372.1 | 2 | 816 | |||
XM_017013884.1 | XP_016869373.1 | 2 | 816 | |||
XM_017013886.1 | XP_016869375.1 | 2 | 816 | |||
GenBank mRNA | AF205075.1 | - | 2 | 725 | ||
AK096879.1 | - | 2 | 816 | |||
AK299048.1 | - | 2 | 772 | |||
AK307567.1 | - | 2 | 774 | |||
AK308488.1 | - | 2 | 718 | |||
BC137424.1 | - | 1 | 611 | 9021422 | ||
BC139755.1 | - | 1 | 151 | 40118841 | ||
BC144618.1 | - | 1 | 611 |