Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030971.3 | NP_112233.2 | 4 | 682 | ||
GenBank mRNA | AK074707.1 | - | 3 | 295 | ||
AK299405.1 | - | 3 | 289 | |||
BC000124.1 | - | 3 | 288 | 3352015 | ||
BG775086.1 | - | 2 | 158 | |||
BI819328.1 | - | 4 | 620 |