Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318758.1 | NP_001305687.1 | 2 | 271 | ||
NM_002504.5 | NP_002495.2 | 2 | 271 | |||
NM_147134.3 | NP_667345.1 | 2 | 271 | |||
GenBank mRNA | AF332009.1 | - | 2 | 176 | ||
AK225750.1 | - | 2 | 144 | |||
AK291643.1 | - | 2 | 161 | |||
AL832342.1 | - | 2 | 153 | |||
BC012151.1 | - | 2 | 141 | 4558442 | ||
BP871782.1 | - | 2 | 213 | |||
DA093124.1 | - | 2 | 269 | |||
DA290058.1 | - | 2 | 217 | |||
DB076795.1 | - | 2 | 271 |