Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030907.3 | NP_112169.2 | 5 | 1277 | ||
GenBank mRNA | BC002946.2 | - | 5 | 1064 | 3955163 | |
BC008702.1 | - | 5 | 1064 | 2966866 |