Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160223.1 | NP_001153695.1 | 3 | 667 | ||
NM_001160224.1 | NP_001153696.1 | 3 | 437 | |||
NM_001160225.1 | NP_001153697.1 | 3 | 316 | |||
NM_030954.3 | NP_112216.3 | 3 | 437 | |||
NR_027668.1 | 3 | 667 | ||||
NR_027669.1 | 3 | 437 | ||||
XM_006716404.2 | XP_006716467.1 | 4 | 424 | |||
XM_011544666.2 | XP_011542968.1 | 3 | 221 | |||
XM_017013881.1 | XP_016869370.1 | 4 | 364 | |||
GenBank mRNA | AK027748.1 | - | 3 | 353 | ||
AK090864.1 | - | 3 | 600 | |||
AK225086.1 | - | 3 | 317 | |||
AK225568.1 | - | 3 | 333 | |||
AL136620.1 | - | 3 | 321 | |||
BC013422.1 | - | 3 | 302 | |||
BC032393.1 | - | 3 | 299 | 5200847 | ||
BC039461.1 | - | 3 | 424 | |||
BC044566.1 | - | 3 | 437 | |||
BC058289.1 | - | 3 | 300 | |||
BG910500.1 | - | 3 | 658 | |||
DA751961.1 | - | 3 | 437 |