Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031212.3 | NP_112489.3 | 4 | 1044 | ||
XM_005270208.1 | XP_005270265.1 | 4 | 1034 | |||
XM_005270211.4 | XP_005270268.1 | 4 | 648 | |||
XM_006718005.3 | XP_006718068.1 | 4 | 708 | |||
XM_011540239.2 | XP_011538541.1 | 4 | 930 | |||
XM_011540240.1 | XP_011538542.1 | 5 | 2180 | |||
XM_017016741.1 | XP_016872230.1 | 4 | 2116 | |||
XM_017016742.1 | XP_016872231.1 | 4 | 1937 | |||
GenBank mRNA | AF267854.1 | - | 1 | 731 | ||
AF327402.1 | - | 4 | 944 | |||
AF327403.1 | - | 4 | 1385 | |||
AJ303077.1 | - | 4 | 944 | |||
AJ303078.1 | - | 4 | 1385 | |||
AK056782.1 | - | 1 | 1084 | |||
AL831943.1 | - | 1 | 606 | |||
BC047312.1 | - | 4 | 991 | |||
BC058937.1 | - | 4 | 953 | 6139015 | ||
BC076399.1 | - | 4 | 984 | 5728224 | ||
BC094821.1 | - | 4 | 727 |