Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005221.5 | NP_005212.1 | 2 | 680 | ||
XM_005250185.3 | XP_005250242.1 | 2 | 1791 | |||
XM_017011803.1 | XP_016867292.1 | 2 | 181 | |||
GenBank mRNA | AK023493.1 | - | 2 | 680 | ||
AK297614.1 | - | 2 | 800 | |||
BC006226.2 | - | 2 | 666 | 3941691 | ||
BT006903.1 | - | 2 | 472 |