Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160213.1 | NP_001153685.1 | 5 | 541 | ||
NM_031293.2 | NP_112583.2 | 4 | 363 | |||
XM_011523357.2 | XP_011521659.1 | 4 | 453 | |||
XM_011523358.2 | XP_011521660.1 | 4 | 393 | |||
XM_011523360.2 | XP_011521662.1 | 4 | 416 | |||
XM_011523361.2 | XP_011521663.1 | 4 | 891 | |||
XM_011523362.2 | XP_011521664.1 | 3 | 198 | |||
GenBank mRNA | AK093285.1 | - | 4 | 359 | ||
AK122925.1 | - | 5 | 541 | |||
AK315004.1 | - | 4 | 344 | |||
AL136795.1 | - | 4 | 323 | |||
BC028423.2 | - | 4 | 350 | 4822328 | ||
DB019901.1 | - | 4 | 453 | |||
DB082449.1 | - | 4 | 395 |