Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031296.2 | NP_112586.1 | 2 | 1280 | ||
XM_011532299.1 | XP_011530601.1 | 3 | 1376 | |||
GenBank mRNA | AF350420.1 | - | 2 | 1002 | ||
AL136904.1 | - | 2 | 1280 | |||
BC036064.1 | - | 2 | 912 | 5288066 | ||
BC111977.1 | - | 2 | 932 | 8327445 |