Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289177.1 | NP_001276106.1 | 9 | 1390 | ||
NM_001289178.1 | NP_001276107.1 | 9 | 1336 | |||
NM_001289179.1 | NP_001276108.1 | 9 | 1381 | |||
NM_001289180.1 | NP_001276109.1 | 9 | 1100 | |||
NM_001321384.1 | NP_001308313.1 | 9 | 1371 | |||
NM_030924.4 | NP_112186.3 | 9 | 1381 | |||
XM_005259653.3 | XP_005259710.1 | 9 | 1363 | |||
XM_011528329.1 | XP_011526631.1 | 6 | 807 | |||
XM_017027332.1 | XP_016882821.1 | 9 | 1208 | |||
XM_017027333.1 | XP_016882822.1 | 9 | 1363 | |||
XM_017027334.1 | XP_016882823.1 | 8 | 1195 | |||
GenBank mRNA | AJ577571.1 | - | 9 | 1284 | ||
AK057412.1 | - | 4 | 592 | |||
AK093433.1 | - | 9 | 1349 | |||
AK225862.1 | - | 8 | 1089 | |||
AK302604.1 | - | 9 | 1338 | |||
AL136854.1 | - | 9 | 1122 | |||
AY009107.1 | - | 9 | 1093 | |||
AY358766.1 | - | 9 | 1381 | |||
BC022027.1 | - | 9 | 1312 | 4825681 | ||
BC090046.1 | - | 9 | 1297 | 30388900 |