Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320153.1 | NP_001307082.1 | 12 | 2453 | ||
NM_001320155.1 | NP_001307084.1 | 12 | 1743 | |||
NM_030937.4 | NP_112199.2 | 11 | 1729 | |||
NR_135154.1 | 13 | 3883 | ||||
XM_011542216.2 | XP_011540518.1 | 11 | 1804 | |||
XM_011542219.2 | XP_011540521.1 | 11 | 1613 | |||
XM_011542221.2 | XP_011540523.1 | 10 | 1444 | |||
XM_017002420.1 | XP_016857909.1 | 12 | 3234 | |||
XM_017002421.1 | XP_016857910.1 | 13 | 2117 | |||
XM_017002422.1 | XP_016857911.1 | 12 | 1851 | |||
GenBank mRNA | AF087903.1 | - | 4 | 773 | ||
AF251294.1 | - | 12 | 2332 | |||
AK000685.1 | - | 8 | 1406 | |||
AK026796.1 | - | 7 | 2026 | |||
AK027770.1 | - | 6 | 2698 | |||
AK056120.1 | - | 12 | 2425 | |||
AK057133.1 | - | 10 | 1444 | |||
AK074112.1 | - | 13 | 3854 | |||
AK225758.1 | - | 11 | 1724 | |||
AK292268.1 | - | 6 | 1252 | |||
AL834432.1 | - | 3 | 599 | |||
AY037150.1 | - | 11 | 1703 | |||
BC071622.1 | - | 6 | 1116 |