Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030945.3 | NP_112207.1 | 6 | 1563 | ||
NM_181435.5 | NP_852100.3 | 6 | 1782 | |||
GenBank mRNA | AF173888.1 | - | 6 | 1465 | ||
AF326976.1 | - | 6 | 1498 | |||
AF329837.1 | - | 6 | 1505 | |||
AK075533.1 | - | 6 | 1510 | |||
AK127389.1 | - | 2 | 1351 | |||
AY358388.1 | - | 6 | 1498 | |||
BC016021.1 | - | 3 | 1199 | |||
BC112925.1 | - | 6 | 1437 | 5199982 | ||
BX640995.1 | - | 7 | 1385 |