Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324511.1 | NP_001311440.1 | 2 | 644 | ||
NM_001324512.1 | NP_001311441.1 | 2 | 644 | |||
NM_030955.3 | NP_112217.2 | 2 | 644 | |||
XM_017009905.1 | XP_016865394.1 | 2 | 632 | |||
GenBank mRNA | AJ250725.1 | - | 2 | 634 | ||
AY358745.1 | - | 2 | 427 | |||
BC058841.1 | - | 2 | 475 | |||
BC131733.1 | - | 2 | 393 | 40124828 | ||
BC139900.1 | - | 2 | 315 | |||
BG108002.1 | - | 2 | 521 |