Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005335.5 | NP_005326.2 | 7 | 632 | ||
GenBank mRNA | AK124015.1 | - | 2 | 1306 | ||
AK225633.1 | - | 6 | 505 | |||
AK298663.1 | - | 6 | 556 | |||
AK312750.1 | - | 7 | 595 | |||
BC016758.2 | - | 7 | 552 | 2966303 | ||
BT006824.1 | - | 6 | 503 | |||
BY994733.1 | - | 3 | 144 | |||
CR456794.1 | - | 6 | 503 | |||
X16663.1 | - | 7 | 545 |