Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267864.1 | NP_001254793.1 | 4 | 329 | ||
NM_031210.5 | NP_112487.1 | 3, 4 | 313 | |||
NR_052025.1 | 4, 5 | 424 | ||||
GenBank mRNA | AF253980.1 | - | 3, 4 | 284 | ||
AF271779.1 | - | 3, 4 | 308 | |||
AJ272055.1 | - | 3, 4 | 272 | |||
AY860853.1 | - | 3, 4 | 272 | |||
BC017895.1 | - | 3, 4 | 281 | 4250073 | ||
BM685909.1 | - | 4, 5 | 389 | |||
BP289922.1 | - | 4 | 301 | |||
BQ083727.1 | - | 4 | 320 | |||
BU944910.1 | - | 3, 4 | 313 | |||
BU960115.1 | - | 3, 4 | 291 | |||
EL951262.1 | - | 3, 4 | 273 |