Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002137.3 | NP_002128.1 | 11 | 1327 | ||
NM_031243.2 | NP_112533.1 | 12 | 1363 | |||
GenBank mRNA | AK129839.1 | - | 11 | 1281 | ||
AK289429.1 | - | 12 | 1316 | |||
BC000506.2 | - | 10 | 1095 | 2822109 | ||
BX537494.1 | - | 9 | 4316 | |||
BX538099.1 | - | 1 | 262 | |||
BX641084.1 | - | 10 | 4901 | |||
M29064.1 | - | 12 | 1296 | |||
M29065.1 | - | 11 | 1266 |