Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031246.3 | NP_112536.2 | 5, 6 | 1228 | ||
GenBank mRNA | BC022316.1 | - | 5, 6 | 1155 | 4731250 | |
M20882.1 | - | 5, 6 | 1139 | |||
M37398.1 | - | 2, 3 | 329 |