Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004499.3 | NP_004490.2 | 5 | 851 | ||
NM_031266.2 | NP_112556.2 | 5 | 851 | |||
GenBank mRNA | AK054600.1 | - | 4 | 899 | ||
AK097657.1 | - | 5 | 852 | |||
AK223425.1 | - | 5 | 828 | |||
BC001616.1 | - | 5 | 810 | 3547193 | ||
BC002625.2 | - | 5 | 810 | 3606731 | ||
BC004561.2 | - | 5 | 818 | 3960088 | ||
BC009359.2 | - | 5 | 814 | 4301401 | ||
BC036708.2 | - | 5 | 777 | 4331453 | ||
M65028.1 | - | 5 | 733 | |||
U76713.1 | - | 4 | 606 | |||
Z36844.1 | - | 1 | 19 |