Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168320.1 | NP_001161792.1 | 8 | 3757 | ||
NM_006675.4 | NP_006666.1 | 9 | 3824 | |||
GenBank mRNA | AF217967.1 | - | 1 | 1464 | ||
AY523992.1 | - | 1 | 2587 | |||
BC034280.1 | - | 1 | 1197 |