Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098614.2 | NP_001092084.1 | 2 | 736 | ||
NM_001098615.1 | NP_001092085.1 | 2 | 1136 | |||
NM_031292.4 | NP_112582.3 | 2 | 751 | |||
XM_006719623.2 | XP_006719686.1 | 2 | 751 | |||
XM_011538790.2 | XP_011537092.1 | 2 | 738 | |||
XM_011538791.2 | XP_011537093.1 | 2 | 751 | |||
XM_017019996.1 | XP_016875485.1 | 2 | 751 | |||
XM_017019997.1 | XP_016875486.1 | 2 | 749 | |||
XM_017019998.1 | XP_016875487.1 | 2 | 1046 | |||
GenBank mRNA | AK092030.1 | - | 2 | 724 | ||
AK225903.1 | - | 2 | 722 | |||
AL136759.1 | - | 2 | 681 | |||
BC018000.2 | - | 1 | 664 | |||
BC020781.1 | - | 2 | 701 | |||
BC033621.2 | - | 1 | 664 | 5195232 | ||
BC068502.1 | - | 2 | 720 | 5259952 | ||
BX647494.1 | - | 2 | 1135 | |||
CR627435.1 | - | 2 | 736 |