Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031296.2 | NP_112586.1 | 2 | 2822 | ||
XM_011532299.1 | XP_011530601.1 | 3 | 2918 | |||
GenBank mRNA | AF350420.1 | - | 2 | 2542 | ||
AL136904.1 | - | 2 | 2822 | |||
BC036064.1 | - | 2 | 2454 | 5288066 |