Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004222.2 | - | 2 | 1206 | 3532660 | |
BC012800.2 | - | 1 | 1222 | 2961135 |