Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTBDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080997.2 | NP_001074466.1 | 6 | 1005 | ||
NM_001270440.1 | NP_001257369.1 | 7 | 2011 | |||
NM_001270441.1 | NP_001257370.1 | 6 | 958 | |||
NM_001270442.1 | NP_001257371.1 | 6 | 946 | |||
NM_001270443.1 | NP_001257372.1 | 6 | 934 | |||
NM_001270444.1 | NP_001257373.1 | 6 | 922 | |||
NM_001270445.1 | NP_001257374.1 | 6 | 916 | |||
NM_031429.2 | NP_113617.1 | 7 | 1278 | |||
XM_005260089.1 | XP_005260146.1 | 7 | 1353 | |||
XM_005260090.3 | XP_005260147.1 | 6 | 1216 | |||
XM_011528343.1 | XP_011526645.1 | 6 | 1019 | |||
XM_011528344.2 | XP_011526646.1 | 6 | 1069 | |||
XM_011528345.1 | XP_011526647.1 | 6 | 1014 | |||
XM_017027343.1 | XP_016882832.1 | 7 | 1308 | |||
XM_017027344.1 | XP_016882833.1 | 7 | 1260 | |||
GenBank mRNA | AB593122.1 | - | 6 | 958 | ||
AK093672.1 | - | 7 | 2011 | |||
AY028917.1 | - | 6 | 982 | |||
BC005063.1 | - | 7 | 1266 | 3344818 |