Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031430.2 | NP_113618.2 | 5 | 1040 | ||
XM_005256811.1 | XP_005256868.1 | 5 | 1019 | |||
GenBank mRNA | AF370391.1 | - | 5 | 977 | ||
AJ278711.1 | - | 5 | 1040 | |||
AJ404317.1 | - | 5 | 1010 | |||
AK055755.1 | - | 5 | 1019 | |||
AK314767.1 | - | 5 | 1019 | |||
BC004961.1 | - | 5 | 589 | |||
BC031621.1 | - | 5 | 834 | 5174974 |