Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040177.2 | NP_001035267.1 | 2 | 342 | ||
NM_001271021.1 | NP_001257950.1 | 2 | 342 | |||
NM_001271025.1 | NP_001257954.1 | 2 | 342 | |||
NR_073125.1 | 2 | 342 | ||||
NR_073126.1 | 2 | 256 | ||||
NR_073127.1 | 2 | 342 | ||||
XM_011519715.2 | XP_011518017.1 | 2 | 639 | |||
XM_011519718.2 | XP_011518020.1 | 2 | 639 | |||
XM_011519719.2 | XP_011518021.1 | 2 | 639 | |||
XM_011519720.2 | XP_011518022.1 | 2 | 638 | |||
XM_011519722.2 | XP_011518024.1 | 2 | 639 | |||
XM_011519724.2 | XP_011518026.1 | 2 | 638 | |||
XM_011519725.2 | XP_011518027.1 | 2 | 636 | |||
XM_011519729.2 | XP_011518031.1 | 2 | 636 | |||
XM_017016743.1 | XP_016872232.1 | 2 | 640 | |||
XM_017016744.1 | XP_016872233.1 | 2 | 639 | |||
XM_017016745.1 | XP_016872234.1 | 2 | 425 | |||
GenBank mRNA | AB040820.1 | - | 2 | 223 | ||
AB040821.1 | - | 2 | 223 | |||
AB040822.1 | - | 2 | 223 | |||
AB055603.1 | - | 2 | 194 | |||
AF263242.1 | - | 2 | 214 | |||
AK074211.1 | - | 2 | 256 | |||
AK302155.1 | - | 2 | 264 | |||
BC002862.2 | - | 2 | 285 | 3941289 | ||
DA006578.1 | - | 2 | 309 |