Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005523.5 | NP_005514.1 | 2 | 2122 | ||
GenBank mRNA | BC040948.1 | - | 2 | 2122 | 5587615 |