Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031457.1 | NP_113645.1 | 2 | 293 | ||
GenBank mRNA | AF237905.1 | - | 2 | 293 | ||
AF350504.1 | - | 2 | 135 | |||
AK310872.1 | - | 2 | 120 | |||
AK310884.1 | - | 2 | 285 | |||
AK310969.1 | - | 2 | 282 | |||
AK315901.1 | - | 2 | 255 | |||
BC022895.1 | - | 2 | 218 | 4691480 |