Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001245002.1 | NP_001231931.1 | 11 | 1774 | ||
NM_001245004.1 | NP_001231933.1 | 10 | 1620 | |||
NM_001245005.1 | NP_001231934.1 | 10 | 1643 | |||
NM_005597.3 | NP_005588.2 | 9 | 1534 | |||
NM_205843.2 | NP_995315.1 | 11 | 1797 | |||
XM_005259564.3 | XP_005259621.1 | 10 | 1696 | |||
XM_017026836.1 | XP_016882325.1 | 9 | 1470 | |||
GenBank mRNA | AK289885.1 | - | 11 | 1710 | ||
AK304816.1 | - | 10 | 1559 | |||
AL042511.1 | - | 4 | 553 | |||
BC012120.1 | - | 9 | 1526 | 4554661 | ||
BI253961.1 | - | 1 | 39 | |||
X92857.1 | - | 9 | 1488 |