Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB049649.1 | - | 3 | 676 | ||
AF161401.1 | - | 3 | 617 | |||
BC013147.1 | - | 3 | 636 | 4340082 |