Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031300.3 | NP_112590.1 | 6 | 1207 | ||
GenBank mRNA | AF114834.1 | - | 6 | 802 | ||
AK222579.1 | - | 6 | 812 | |||
AK303397.1 | - | 5 | 891 | |||
AK316468.1 | - | 5 | 888 | |||
AL833959.1 | - | 3 | 1853 | |||
BC000745.2 | - | 6 | 796 | 2821596 | ||
BC032586.1 | - | 5 | 888 |